Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Inhibits the poly(ADP-ribosyl)ation activity of PARP1 and the degradation of PARP1 by CASP3 following genotoxic stress. Binds to double-stranded RNA or DNA without sequence specificity. Involved in development of the eye and of primordial germ cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV X-transactivated gene 5 protein; HBV XAg-transactivated protein 5; HLA-HA8; Minor histocompatibility antigen HA-8; penguin homolog; protein 5 transactivated by hepatitis B virus X antigen (HBxAg); Pumilio homolog 3; pumilio RNA-binding family member 3
Gene Aliases: cPERP-C; HA-8; HLA-HA8; KIAA0020; PEN; PUF-A; PUF6; PUM3; XTP5
UniProt ID: (Human) Q5SZY9
Entrez Gene ID: (Human) 9933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support