Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEVKGKKQFT GKNTKTAQEK NRFHKNSDSG SSKTFPTRKV AKEGGPKVTS RNFEKSITKL GKKGVKQFKN KQQGDKSPKN KFQPANKFNK KRKFQPDGRS DESAAKKPKW DDFKKKKKEL KQSRQLSDKT NYDIVVRAKQ MWEILRRKDC DKEKRVKLMS DLQKLIQGKI KTIAFAHDST RVIQCYIQYG NEEQRKQAFE ELRDDLVELS KAKYSRNIVK KFLMYGSKPQ IAEIIRSFKG HVRKMLRHAE ASAIVEYAYN DKAILEQRNM LTEELYGNTF QLYKSADHPT LDKVLELQPE KLELIMDEMK QILTPMAQKE AVIKHSLVHK VFLDFFTYAP PKLRSEMIEA IREAVVYLAH THDGARVAMH CLWHGTPKDR KVIVKTMKTY VEKVANGQYS HLVLLAAFDC IDDTKLVKQI IISEIISSLP SIVNDKYGRK VLLYLLSPRD PAHTVREIIE VLQKGDGNAH SKKDTEVRRR ELLESISPAL LSYLQEHAQE VVLDKSACVL VSDILGSATG DVQPTMNAIA SLAATGLHPG GKDGELHIAE HPAGHLVLKW LIEQDKKMKE NGREGCFAKT LVEHVGMKNL KSWASVNRGA IILSSLLQSC DLEVANKVKA ALKSLIPTLE KTKSTSKGIE ILLEKLST
Inhibits the poly(ADP-ribosyl)ation activity of PARP1 and the degradation of PARP1 by CASP3 following genotoxic stress. Binds to double-stranded RNA or DNA without sequence specificity. Involved in development of the eye and of primordial germ cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV X-transactivated gene 5 protein; HBV XAg-transactivated protein 5; HLA-HA8; Minor histocompatibility antigen HA-8; penguin homolog; protein 5 transactivated by hepatitis B virus X antigen (HBxAg); Pumilio homolog 3; pumilio RNA-binding family member 3
Gene Aliases: cPERP-C; HA-8; HLA-HA8; KIAA0020; PEN; PUF-A; PUF6; PUM3; XTP5
UniProt ID: (Human) Q5SZY9
Entrez Gene ID: (Human) 9933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support