Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MRSKPDKDLL MEEDDMALAN VVMQEAQKKL ISQVQKRNFI ENIIPIIISL KTVLEKNKIP ALRELMHYLR EVMQDYRDEL KDFFAVDKQL ASELEYDMKK YQEQLVQEQE LAKHADVAGT AGGAEVAPVA QVALCLETVP VPAGQENPAM SPAVSQPCTP RASAGHVAVS SPTPETGPLQ RLLPKARPMS LSTIAILNSV KKAVESKSRH RSRSLGVLPF TLNSGSPEKT CSQVSSYSLE QESNGEIEHV TKRAI
This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Condensin-2 complex subunit D3; hCAP-D3; Non-SMC condensin II complex subunit D3; non-SMC condensin II complex, subunit D3
Gene Aliases: CAP-D3; CAPD3; hCAP-D3; hcp-6; hHCP-6; KIAA0056; NCAPD3
UniProt ID: (Human) P42695
Entrez Gene ID: (Human) 23310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support