Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLFDPAAWLH GPPETSGPPD GQRRHRPESG SGSREAPTSC ESLDVSPPGA REEPWLKELS LAFLQQYVQY LQSIGFVLVP LRPPSPARST SRPRAMAILG TEGRGSFSCP KTKTDGSPKS TSSPVTTYHL QRALPGGIIL MELAFQGCYF CVKQFALECS RIPMGQAVNS QLSMLFTEEC DKVRDLMHVH SFSYDFHLRL VHQHVLGAHL VLRHGYHLTT FLRHFLAHHP DGPHFGRNHI YQGTLELPTP LIAAHQLYNY VADHASSYHM KPLRMARPGG PEHNEYALVS AWHSSGSYLD SEGLRHQDDF DVSLLVCHCA APFEEQGEAE RHVLRLQFFV VLTSQRELFP RLTADMRRFR KPPRLPPEPE APGSSAGSPG EASGLILAPG PAPLFPPLAA EVGMARARLA QLVRLAGGHC RRDTLWKRLF LLEPPGPDRL RLGGRLALAE LEELLEAVHA KSIGDIDPQL DCFLSMTVSW YQSLIKVLLS RFPQSCRHFQ SPDLGTQYLV VLNQKFTDCF VLVFLDSHLG KTSLTVVFRE PFPVQPQDSE SPPAQLVSTY HHLESVINTA CFTLWTRLL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KICSTOR complex protein SZT2; RP11-506B15.1; RP11-506B15.3; Seizure threshold 2 protein homolog
Gene Aliases: C1orf84; EIEE18; KIAA0467; SZT2; SZT2A; SZT2B
UniProt ID: (Human) Q5T011
Entrez Gene ID: (Human) 23334
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support