Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. For mouse tissues: 1:80 - 1:320. Epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis and Orangutan
C2CD5 gene ontology annotations related to this gene include calcium-dependent phospholipid binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 138 kDa C2 domain-containing phosphoprotein; C2 calcium-dependent domain containing 5; C2 domain-containing phosphoprotein of 138 kDa; C2 domain-containing protein 5
Gene Aliases: 5730419I09Rik; C030008B15Rik; C2CD5; CDP138; KIAA0528
UniProt ID: (Human) Q86YS7, (Mouse) Q7TPS5
Entrez Gene ID: (Human) 9847, (Mouse) 74741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support