Product References
Hyaluronan in articular cartilage: Analysis of hip osteoarthritis and osteonecrosis of femoral head.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Zhang J,Nishida Y,Koike H,Ito K,Zhuo L,Nishida K,Kimata K,Ikuta K,Sakai T,Urakawa H,Seki T,Imagama S
Wed Feb 01 00:00:00 EST 2023
KIAA1199 promotes oxaliplatin resistance and epithelial mesenchymal transition of colorectal cancer via protein O-GlcNAcylation.
Translational oncology
Hua Q,Lu Y,Wang D,Da J,Peng W,Sun G,Gu K,Wang H,Zhu Y
Wed Feb 01 00:00:00 EST 2023
KIAA1199 promotes oxaliplatin resistance and epithelial mesenchymal transition of colorectal cancer via protein O-GlcNAcylation.
Translational oncology
Hua Q,Lu Y,Wang D,Da J,Peng W,Sun G,Gu K,Wang H,Zhu Y
Wed Feb 01 00:00:00 EST 2023
Downregulation of CEMIP enhances radiosensitivity by promoting DNA damage and apoptosis in colorectal cancer.
Medical oncology (Northwood, London, England)
Weng J,Zhang Y,Liang W,Xie Y,Wang K,Xu Q,Ding Y,Li Y
Fri Jan 06 00:00:00 EST 2023
CEMIP, acting as a scaffold protein for bridging GRAF1 and MIB1, promotes colorectal cancer metastasis via activating CDC42/MAPK pathway.
Cell death & disease
Xu G,Zhao L,Hua Q,Wang L,Liu H,Lin Z,Jin M,Wang J,Zhou P,Yang K,Wu G,Yu D,Zhang D,Zhang T
Mon Feb 27 00:00:00 EST 2023
CEMIP, acting as a scaffold protein for bridging GRAF1 and MIB1, promotes colorectal cancer metastasis via activating CDC42/MAPK pathway.
Cell death & disease
Xu G,Zhao L,Hua Q,Wang L,Liu H,Lin Z,Jin M,Wang J,Zhou P,Yang K,Wu G,Yu D,Zhang D,Zhang T
Mon Feb 27 00:00:00 EST 2023
CEMIP, acting as a scaffold protein for bridging GRAF1 and MIB1, promotes colorectal cancer metastasis via activating CDC42/MAPK pathway.
Cell death & disease
Xu G,Zhao L,Hua Q,Wang L,Liu H,Lin Z,Jin M,Wang J,Zhou P,Yang K,Wu G,Yu D,Zhang D,Zhang T
Mon Feb 27 00:00:00 EST 2023
CEMIP, acting as a scaffold protein for bridging GRAF1 and MIB1, promotes colorectal cancer metastasis via activating CDC42/MAPK pathway.
Cell death & disease
Xu G,Zhao L,Hua Q,Wang L,Liu H,Lin Z,Jin M,Wang J,Zhou P,Yang K,Wu G,Yu D,Zhang D,Zhang T
Mon Feb 27 00:00:00 EST 2023
KIAA1199 deficiency enhances skeletal stem cell differentiation to osteoblasts and promotes bone regeneration.
Nature communications
Chen L,Shi K,Ditzel N,Qiu W,Figeac F,Nielsen LHD,Tencerova M,Kowal JM,Ding M,Andreasen CM,Andersen TL,Kassem M
Mon Apr 10 00:00:00 EDT 2023
A top-down approach to uncover the hidden ligandome of low-density lipoprotein receptor-related protein 1 in cartilage.
Matrix biology : journal of the International Society for Matrix Biology
Yamamoto K,Scavenius C,Meschis MM,Gremida AME,Mogensen EH,Thøgersen IB,Bonelli S,Scilabra SD,Jensen A,Santamaria S,Ahnström J,Bou-Gharios G,Enghild JJ,Nagase H
Thu Sep 01 00:00:00 EDT 2022
A top-down approach to uncover the hidden ligandome of low-density lipoprotein receptor-related protein 1 in cartilage.
Matrix biology : journal of the International Society for Matrix Biology
Yamamoto K,Scavenius C,Meschis MM,Gremida AME,Mogensen EH,Thøgersen IB,Bonelli S,Scilabra SD,Jensen A,Santamaria S,Ahnström J,Bou-Gharios G,Enghild JJ,Nagase H
Thu Sep 01 00:00:00 EDT 2022
A top-down approach to uncover the hidden ligandome of low-density lipoprotein receptor-related protein 1 in cartilage.
Matrix biology : journal of the International Society for Matrix Biology
Yamamoto K,Scavenius C,Meschis MM,Gremida AME,Mogensen EH,Thøgersen IB,Bonelli S,Scilabra SD,Jensen A,Santamaria S,Ahnström J,Bou-Gharios G,Enghild JJ,Nagase H
Thu Sep 01 00:00:00 EDT 2022
A top-down approach to uncover the hidden ligandome of low-density lipoprotein receptor-related protein 1 in cartilage.
Matrix biology : journal of the International Society for Matrix Biology
Yamamoto K,Scavenius C,Meschis MM,Gremida AME,Mogensen EH,Thøgersen IB,Bonelli S,Scilabra SD,Jensen A,Santamaria S,Ahnström J,Bou-Gharios G,Enghild JJ,Nagase H
Thu Sep 01 00:00:00 EDT 2022
Expression and regulation of recently discovered hyaluronidases, HYBID and TMEM2, in chondrocytes from knee osteoarthritic cartilage.
Scientific reports
Shiozawa J,de Vega S,Yoshinaga C,Ji X,Negishi Y,Momoeda M,Nakamura T,Yoshida H,Kaneko H,Ishijima M,Okada Y
Fri Oct 14 00:00:00 EDT 2022
Expression and regulation of recently discovered hyaluronidases, HYBID and TMEM2, in chondrocytes from knee osteoarthritic cartilage.
Scientific reports
Shiozawa J,de Vega S,Yoshinaga C,Ji X,Negishi Y,Momoeda M,Nakamura T,Yoshida H,Kaneko H,Ishijima M,Okada Y
Fri Oct 14 00:00:00 EDT 2022
KIAA1199 Correlates With Tumor Microenvironment and Immune Infiltration in Lung Adenocarcinoma as a Potential Prognostic Biomarker.
Pathology oncology research : POR
Shen X,Mo X,Tan W,Mo X,Li L,Yu F,He J,Deng Z,Xing S,Chen Z,Yang J
Thu Dec 01 00:00:00 EST 2022
CEMIP Promotes Osteosarcoma Progression and Metastasis Through Activating Notch Signaling Pathway.
Frontiers in oncology
Cheng J,Zhang Y,Wan R,Zhou J,Wu X,Fan Q,He J,Tan W,Deng Y
Sat Aug 13 00:00:00 EDT 2022
CEMIP Promotes Osteosarcoma Progression and Metastasis Through Activating Notch Signaling Pathway.
Frontiers in oncology
Cheng J,Zhang Y,Wan R,Zhou J,Wu X,Fan Q,He J,Tan W,Deng Y
Sat Aug 13 00:00:00 EDT 2022
CEMIP Promotes Osteosarcoma Progression and Metastasis Through Activating Notch Signaling Pathway.
Frontiers in oncology
Cheng J,Zhang Y,Wan R,Zhou J,Wu X,Fan Q,He J,Tan W,Deng Y
Sat Aug 13 00:00:00 EDT 2022
CEMIP Promotes Osteosarcoma Progression and Metastasis Through Activating Notch Signaling Pathway.
Frontiers in oncology
Cheng J,Zhang Y,Wan R,Zhou J,Wu X,Fan Q,He J,Tan W,Deng Y
Sat Aug 13 00:00:00 EDT 2022
Single-cell analysis of human basal cell carcinoma reveals novel regulators of tumor growth and the tumor microenvironment.
Science advances
Guerrero-Juarez CF,Lee GH,Liu Y,Wang S,Karikomi M,Sha Y,Chow RY,Nguyen TTL,Iglesias VS,Aasi S,Drummond ML,Nie Q,Sarin K,Atwood SX
Fri Jun 10 00:00:00 EDT 2022
Identification of metastasis-associated exoDEPs in colorectal cancer using label-free proteomics.
Translational oncology
Liu X,Li N,Zhang C,Wu X,Zhang S,Dong G,Liu G
Sun May 01 00:00:00 EDT 2022
Identification of metastasis-associated exoDEPs in colorectal cancer using label-free proteomics.
Translational oncology
Liu X,Li N,Zhang C,Wu X,Zhang S,Dong G,Liu G
Sun May 01 00:00:00 EDT 2022
Altered regulation of DPF3, a member of the SWI/SNF complexes, underlies the 14q24 renal cancer susceptibility locus.
American journal of human genetics
Colli LM,Jessop L,Myers TA,Camp SY,Machiela MJ,Choi J,Cunha R,Onabajo O,Mills GC,Schmid V,Brodie SA,Delattre O,Mole DR,Purdue MP,Yu K,Brown KM,Chanock SJ
Thu Sep 02 00:00:00 EDT 2021
Lenvatinib induces anticancer activity in gallbladder cancer by targeting AKT.
Journal of Cancer
Ye J,Qi L,Liang J,Zong K,Liu W,Li R,Feng R,Zhai W
Tue May 18 00:00:00 EDT 2021
Hypoxia increases KIAA1199/CEMIP expression and enhances cell migration in pancreatic cancer.
Scientific reports
Oba T,Sato N,Adachi Y,Amaike T,Kudo Y,Koga A,Kohi S,Hirata K
Tue Sep 14 00:00:00 EDT 2021
Hypoxia increases KIAA1199/CEMIP expression and enhances cell migration in pancreatic cancer.
Scientific reports
Oba T,Sato N,Adachi Y,Amaike T,Kudo Y,Koga A,Kohi S,Hirata K
Tue Sep 14 00:00:00 EDT 2021
Efficacy of hyaluronic acid on intervertebral disc inflammation: An in vitro study using notochordal cell lines and human disc cells.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Yamamoto T,Suzuki S,Fujii T,Mima Y,Watanabe K,Matsumoto M,Nakamura M,Fujita N
Fri Oct 01 00:00:00 EDT 2021
Efficacy of hyaluronic acid on intervertebral disc inflammation: An in vitro study using notochordal cell lines and human disc cells.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Yamamoto T,Suzuki S,Fujii T,Mima Y,Watanabe K,Matsumoto M,Nakamura M,Fujita N
Fri Oct 01 00:00:00 EDT 2021
Deletion of Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization (HYBID) Results in Attenuation of Osteoarthritis in Mice.
The American journal of pathology
Momoeda M,de Vega S,Kaneko H,Yoshinaga C,Shimoda M,Nakamura T,Endo Y,Yoshida H,Kaneko K,Ishijima M,Okada Y
Mon Nov 01 00:00:00 EDT 2021
HYBID derived from tumor cells and tumor-associated macrophages contribute to the glioblastoma growth.
Brain research
Tsuji S,Nakamura S,Yamada T,de Vega S,Okada Y,Inoue S,Shimazawa M,Hara H
Sun Aug 01 00:00:00 EDT 2021
HYBID derived from tumor cells and tumor-associated macrophages contribute to the glioblastoma growth.
Brain research
Tsuji S,Nakamura S,Yamada T,de Vega S,Okada Y,Inoue S,Shimazawa M,Hara H
Sun Aug 01 00:00:00 EDT 2021
Overexpression of KIAA1199, a novel strong hyaluronidase, is a poor prognostic factor in patients with osteosarcoma.
Journal of orthopaedic surgery and research
Ito K,Nishida Y,Ikuta K,Urakawa H,Koike H,Sakai T,Zhang J,Shimoyama Y,Imagama S
Wed Jul 07 00:00:00 EDT 2021
Comprehensive histological investigation of age-related changes in dermal extracellular matrix and muscle fibers in the upper lip vermilion.
International journal of cosmetic science
Gomi T,Imamura T
Sat Aug 01 00:00:00 EDT 2020
Serum KIAA1199 is an advanced-stage prognostic biomarker and metastatic oncogene in cholangiocarcinoma.
Aging
Zhai X,Wang W,Ma Y,Zeng Y,Dou D,Fan H,Song J,Yu X,Xin D,Du G,Jiang Z,Zhang H,Zhang X,Jin B
Tue Nov 10 00:00:00 EST 2020
Serum KIAA1199 is an advanced-stage prognostic biomarker and metastatic oncogene in cholangiocarcinoma.
Aging
Zhai X,Wang W,Ma Y,Zeng Y,Dou D,Fan H,Song J,Yu X,Xin D,Du G,Jiang Z,Zhang H,Zhang X,Jin B
Tue Nov 10 00:00:00 EST 2020
Forced expression of KIAA1199, a novel hyaluronidase, inhibits tumorigenicity of low-grade chondrosarcoma.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Koike H,Nishida Y,Shinomura T,Zhuo L,Hamada S,Ikuta K,Ito K,Kimata K,Ushida T,Ishiguro N
Tue Sep 01 00:00:00 EDT 2020
Co-expression of KIAA1199 and hypoxia-inducible factor 1α is a biomarker for an unfavorable prognosis in hepatocellular carcinoma.
Medicine
Wang D,Lu S,Zhang X,Huang L,Zhao H
Fri Dec 11 00:00:00 EST 2020
Co-expression of KIAA1199 and hypoxia-inducible factor 1α is a biomarker for an unfavorable prognosis in hepatocellular carcinoma.
Medicine
Wang D,Lu S,Zhang X,Huang L,Zhao H
Fri Dec 11 00:00:00 EST 2020
A novel metastatic promoter CEMIP and its downstream molecular targets and signaling pathway of cellular migration and invasion in SCLC cells based on proteome analysis.
Journal of cancer research and clinical oncology
Li L,Pan Y,Mo X,Wei T,Song J,Luo M,Huang G,Teng C,Liang K,Mao N,Yang J
Thu Oct 01 00:00:00 EDT 2020
Implication of HYBID (Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization) in Hyaluronan Degradation by Synovial Fibroblasts in Patients with Knee Osteoarthritis.
The American journal of pathology
Shiozawa J,de Vega S,Cilek MZ,Yoshinaga C,Nakamura T,Kasamatsu S,Yoshida H,Kaneko H,Ishijima M,Kaneko K,Okada Y
Fri May 01 00:00:00 EDT 2020
Implication of HYBID (Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization) in Hyaluronan Degradation by Synovial Fibroblasts in Patients with Knee Osteoarthritis.
The American journal of pathology
Shiozawa J,de Vega S,Cilek MZ,Yoshinaga C,Nakamura T,Kasamatsu S,Yoshida H,Kaneko H,Ishijima M,Kaneko K,Okada Y
Fri May 01 00:00:00 EDT 2020
Implication of HYBID (Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization) in Hyaluronan Degradation by Synovial Fibroblasts in Patients with Knee Osteoarthritis.
The American journal of pathology
Shiozawa J,de Vega S,Cilek MZ,Yoshinaga C,Nakamura T,Kasamatsu S,Yoshida H,Kaneko H,Ishijima M,Kaneko K,Okada Y
Fri May 01 00:00:00 EDT 2020
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 EST 2019
CEMIP (KIAA1199) induces a fibrosis-like process in osteoarthritic chondrocytes.
Cell death & disease
Deroyer C,Charlier E,Neuville S,Malaise O,Gillet P,Kurth W,Chariot A,Malaise M,de Seny D
Mon Feb 04 00:00:00 EST 2019
KIAA1199 is a secreted molecule that enhances osteoblastic stem cell migration and recruitment.
Cell death & disease
Chen L,Shi K,Andersen TL,Qiu W,Kassem M
Tue Feb 12 00:00:00 EST 2019
CEMIP upregulates BiP to promote breast cancer cell survival in hypoxia.
Oncotarget
Banach A,Jiang YP,Roth E,Kuscu C,Cao J,Lin RZ
Tue Jul 02 00:00:00 EDT 2019
KIAA1199 overexpression is associated with abnormal expression of EMT markers and is a novel independent prognostic biomarker for hepatocellular carcinoma.
OncoTargets and therapy
Jiang Z,Zhai X,Shi B,Luo D,Jin B
Wed Mar 30 00:00:00 EDT 2022
KIAA1199 overexpression is associated with abnormal expression of EMT markers and is a novel independent prognostic biomarker for hepatocellular carcinoma.
OncoTargets and therapy
Jiang Z,Zhai X,Shi B,Luo D,Jin B
Wed Mar 30 00:00:00 EDT 2022
KIAA1199 overexpression is associated with abnormal expression of EMT markers and is a novel independent prognostic biomarker for hepatocellular carcinoma.
OncoTargets and therapy
Jiang Z,Zhai X,Shi B,Luo D,Jin B
Wed Mar 30 00:00:00 EDT 2022
KIAA1199 overexpression is associated with abnormal expression of EMT markers and is a novel independent prognostic biomarker for hepatocellular carcinoma.
OncoTargets and therapy
Jiang Z,Zhai X,Shi B,Luo D,Jin B
Wed Mar 30 00:00:00 EDT 2022
Down-regulation of KIAA1199/CEMIP by miR-216a suppresses tumor invasion and metastasis in colorectal cancer.
International journal of cancer
Zhang D,Zhao L,Shen Q,Lv Q,Jin M,Ma H,Nie X,Zheng X,Huang S,Zhou P,Wu G,Zhang T
Mon May 15 00:00:00 EDT 2017
Down-regulation of KIAA1199/CEMIP by miR-216a suppresses tumor invasion and metastasis in colorectal cancer.
International journal of cancer
Zhang D,Zhao L,Shen Q,Lv Q,Jin M,Ma H,Nie X,Zheng X,Huang S,Zhou P,Wu G,Zhang T
Mon May 15 00:00:00 EDT 2017
Down-regulation of KIAA1199/CEMIP by miR-216a suppresses tumor invasion and metastasis in colorectal cancer.
International journal of cancer
Zhang D,Zhao L,Shen Q,Lv Q,Jin M,Ma H,Nie X,Zheng X,Huang S,Zhou P,Wu G,Zhang T
Mon May 15 00:00:00 EDT 2017
Down-regulation of KIAA1199/CEMIP by miR-216a suppresses tumor invasion and metastasis in colorectal cancer.
International journal of cancer
Zhang D,Zhao L,Shen Q,Lv Q,Jin M,Ma H,Nie X,Zheng X,Huang S,Zhou P,Wu G,Zhang T
Mon May 15 00:00:00 EDT 2017
KIAA1199/CEMIP/HYBID overexpression predicts poor prognosis in pancreatic ductal adenocarcinoma.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.]
Koga A,Sato N,Kohi S,Yabuki K,Cheng XB,Hisaoka M,Hirata K
Wed Aug 09 00:00:00 EDT 2017
KIAA1199/CEMIP/HYBID overexpression predicts poor prognosis in pancreatic ductal adenocarcinoma.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.]
Koga A,Sato N,Kohi S,Yabuki K,Cheng XB,Hisaoka M,Hirata K
Wed Aug 09 00:00:00 EDT 2017
KIAA1199 is induced by inflammation and enhances malignant phenotype in pancreatic cancer.
Oncotarget
Kohi S,Sato N,Koga A,Matayoshi N,Hirata K
Tue Mar 07 00:00:00 EST 2017
KIAA1199 is induced by inflammation and enhances malignant phenotype in pancreatic cancer.
Oncotarget
Kohi S,Sato N,Koga A,Matayoshi N,Hirata K
Tue Mar 07 00:00:00 EST 2017
KIAA1199 promotes migration and invasion by Wnt/β-catenin pathway and MMPs mediated EMT progression and serves as a poor prognosis marker in gastric cancer.
PloS one
Jia S,Qu T,Wang X,Feng M,Yang Y,Feng X,Ma R,Li W,Hu Y,Feng Y,Ji K,Li Z,Jiang W,Ji J
Thu Apr 27 00:00:00 EDT 2017
KIAA1199 promotes migration and invasion by Wnt/β-catenin pathway and MMPs mediated EMT progression and serves as a poor prognosis marker in gastric cancer.
PloS one
Jia S,Qu T,Wang X,Feng M,Yang Y,Feng X,Ma R,Li W,Hu Y,Feng Y,Ji K,Li Z,Jiang W,Ji J
Thu Apr 27 00:00:00 EDT 2017
Cochlear Cell Modeling Using Disease-Specific iPSCs Unveils a Degenerative Phenotype and Suggests Treatments for Congenital Progressive Hearing Loss.
Cell reports
Hosoya M,Fujioka M,Sone T,Okamoto S,Akamatsu W,Ukai H,Ueda HR,Ogawa K,Matsunaga T,Okano H
Tue Jan 03 00:00:00 EST 2017
Cochlear Cell Modeling Using Disease-Specific iPSCs Unveils a Degenerative Phenotype and Suggests Treatments for Congenital Progressive Hearing Loss.
Cell reports
Hosoya M,Fujioka M,Sone T,Okamoto S,Akamatsu W,Ukai H,Ueda HR,Ogawa K,Matsunaga T,Okano H
Tue Jan 03 00:00:00 EST 2017
Functional Genomic mRNA Profiling of Colorectal Adenomas: Identification and in vivo Validation of CD44 and Splice Variant CD44v6 as Molecular Imaging Targets.
Theranostics
Hartmans E,Orian-Rousseau V,Matzke-Ogi A,Karrenbeld A,de Groot DJ,de Jong S,van Dam GM,Fehrmann RS,Nagengast WB
Mon Oct 23 00:00:00 EDT 2017
Functional Genomic mRNA Profiling of Colorectal Adenomas: Identification and in vivo Validation of CD44 and Splice Variant CD44v6 as Molecular Imaging Targets.
Theranostics
Hartmans E,Orian-Rousseau V,Matzke-Ogi A,Karrenbeld A,de Groot DJ,de Jong S,van Dam GM,Fehrmann RS,Nagengast WB
Mon Oct 23 00:00:00 EDT 2017
Conditional knockdown of hyaluronidase 2 in articular cartilage stimulates osteoarthritic progression in a mice model.
Scientific reports
Higuchi Y,Nishida Y,Kozawa E,Zhuo L,Arai E,Hamada S,Morita D,Ikuta K,Kimata K,Ushida T,Ishiguro N
Tue Aug 01 00:00:00 EDT 2017
Conditional knockdown of hyaluronidase 2 in articular cartilage stimulates osteoarthritic progression in a mice model.
Scientific reports
Higuchi Y,Nishida Y,Kozawa E,Zhuo L,Arai E,Hamada S,Morita D,Ikuta K,Kimata K,Ushida T,Ishiguro N
Tue Aug 01 00:00:00 EDT 2017
Crohn's Disease Fibroblasts Overproduce the Novel Protein KIAA1199 to Create Proinflammatory Hyaluronan Fragments.
Cellular and molecular gastroenterology and hepatology
Soroosh A,Albeiroti S,West GA,Willard B,Fiocchi C,de la Motte CA
Sun May 01 00:00:00 EDT 2016
Crohn's Disease Fibroblasts Overproduce the Novel Protein KIAA1199 to Create Proinflammatory Hyaluronan Fragments.
Cellular and molecular gastroenterology and hepatology
Soroosh A,Albeiroti S,West GA,Willard B,Fiocchi C,de la Motte CA
Sun May 01 00:00:00 EDT 2016
Distinct Expression Pattern of a Deafness Gene, KIAA1199, in a Primate Cochlea.
BioMed research international
Hosoya M,Fujioka M,Okano H,Ogawa K
Tue Feb 14 00:00:00 EST 2017
Distinct Expression Pattern of a Deafness Gene, KIAA1199, in a Primate Cochlea.
BioMed research international
Hosoya M,Fujioka M,Okano H,Ogawa K
Tue Feb 14 00:00:00 EST 2017
Functional proteomic analysis reveals the involvement of KIAA1199 in breast cancer growth, motility and invasiveness.
BMC cancer
Jami MS,Hou J,Liu M,Varney ML,Hassan H,Dong J,Geng L,Wang J,Yu F,Huang X,Peng H,Fu K,Li Y,Singh RK,Ding SJ
Sat Mar 15 00:00:00 EDT 2014
Functional proteomic analysis reveals the involvement of KIAA1199 in breast cancer growth, motility and invasiveness.
BMC cancer
Jami MS,Hou J,Liu M,Varney ML,Hassan H,Dong J,Geng L,Wang J,Yu F,Huang X,Peng H,Fu K,Li Y,Singh RK,Ding SJ
Sat Mar 15 00:00:00 EDT 2014