Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
KIAA1217 required for normal development of intervertebral disks.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: likely orthologue of Mus musculus enhancer trap locus 4; RP11-324E23.1; Sickle tail protein homolog
Gene Aliases: KIAA1217; SKT
UniProt ID: (Human) Q5T5P2
Entrez Gene ID: (Human) 56243
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support