Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAKEGVEKAE ETEQMIEKEA GKEPAEGGGG DGSHRLGDAQ EMRAVVLAGF GGLNKLRLFR KAMPEPQDGE LKIRVKACGL NFIDLMVRQG NIDNPPKTPL VPGFECSGIV EALGDSVKGY EIGDRVMAFV NYNAWAEVVC TPVEFVYKIP DDMSFSEAAA FPMNFVTAYV MLFEVANLRE GMSVLVHSAG GGVGQAVAQL CSTVPNVTVF GTASTFKHEA IKDSVTHLFD RNADYVQEVK RISAEGVDIV LDCLCGDNTG KGLSLLKPLG TYILYGSSNM VTGETKSFFS FAKSWWQVEK VNPIKLYEEN KVIAGFSLLN LLFKQGRAGL IRGVVEKLIG LYNQKKIKPV VDSLWALEEV KEAMQRIHDR GNIGKLILDV EKTPTPLMAN DSTETSEAGE EEEDHEGDSE NKERMPFIQ
VAT1L is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptic vesicle membrane protein VAT-1 homolog-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases: KIAA1576; VAT1L
UniProt ID: (Human) Q9HCJ6
Entrez Gene ID: (Human) 57687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support