Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Dog: 86%; Human: 100%
KIAA1614 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein KIAA1614
Gene Aliases: KIAA1614
UniProt ID: (Human) Q5VZ46
Entrez Gene ID: (Human) 57710
Molecular Function:
tight junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support