Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSQFKRQRIN PLPGGRNFSG TASTSLLGPP PGLLTPPVAT ELSQNARHLQ GGEKQRVFTG IVTSLHDYFG VVDEEVFFQL SVVKGRLPQL GEKVLVKAAY NPGQAVPWNA VKVQTLSNQP LLKSPAPPLL HVAALGQKQG ILGAQPQLIF QPHRIPPLFP QKPLSLFQTS HTLHLSHLNR FPARGPHGRL DQGRSDDYDS KKRKQRAGGE PWGAKKPRHD LPPYRVHLTP YTVDSPICDF LELQRRYRSL LVPSDFLSVH LSWLSAFPLS QPFSLHHPSR IQVSSEKEAA PDAGAEPITA DSDPAYSSKV LLLSSPGLEE LYRCCMLFVD DMAEPRETPE HPLKQIKFLL GRKEEEAVLV GGEWSPSLDG LDPQADPQVL VRTAIRCAQA QTGIDLSGCT KWWRFAEFQY LQPGPPRRLQ TVVVYLPDVW TIMPTLEEWE ALCQQKAAEA APPTQEAQGE TEPTEQAPDA LEQAADTSRR NAETPEATTQ QETDTDLPEA PPPPLEPAVI ARPGCVNLSL HGIVEDRRPK ERISFEVMVL AELFLEMLQR DFGYRVYKML LSLPEKVVSP PEPEKEEAAK EEATKEEEAI KEEVVKEPKD EAQNEGPATE SEAPLKEDGL LPKPLSSGGE EEEKPRGEAS EDLCEMALDP ELLLLRDDGE EEFAGAKLED SEVRSVASNQ SEMEFSSLQD MPKELDPSAV LPLDCLLAFV FFDANWCGYL HRRDLERILL TLGIRLSAEQ AKQLVSRVVT QNICQYRSLQ YSRQEGLDGG LPEEVLFGNL DLLPPPGKST KPGAAPTEHK ALVSHNGSLI NVGSLLQRAE QQDSGRLYLE NKIHTLELKL EESHNRFSAT EVTNKTLAAE MQELRVRLAE AEETARTAER QKSQLQRLLQ ELRRRLTPLQ LEIQRVVEKA DSWVEKEEPA PSN
DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell cycle and apoptosis regulator protein 2; Cell division cycle and apoptosis regulator protein 2; DBC-1; DBC.1; DBCCR1; DBIRD complex subunit KIAA1967; deleted in breast cancer 1; Deleted in breast cancer gene 1 protein; FAM5A; NET35; p30 DBC; p30 DBC protein
Gene Aliases: CCAR2; DBC-1; DBC1; KIAA1967; NET35; p30 DBC; p30DBC
UniProt ID: (Human) Q8TF34
Entrez Gene ID: (Human) 57805
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support