Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: WSRGEPGAAE PSACLEAATR AWRGLRERGE VVPLGPGVPA LVANGFLALD VAANRLWVTP GEREPAVAPD FVPFVQLRPL SALAEAGEAV LLLREGLLRR VRCLQLGSPG PGPVAAGPGP ASVSGLAAGS GRDCVLLQED FLAHRGRPHV YLQRIQLNNP TERVAALQTV GPTAGPAPKA FTSTLEKVGD HQFLLYSGRS PPTPTGLVHL VVVAAKKLVN RLQVAPKTQL DETVLWVVHV SGPINPQVLK SKAAKELKAL QDLARKEMLE LLDMPAAELL QDHQLLWAQL FSPGVEMKKI TDTHTPSGLT VNLTLYYMLS CSPAPLLSPS LSHRERDQME ST
KIAA2013 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP5-1077B9.1; Uncharacterized protein KIAA2013
Gene Aliases: KIAA2013
UniProt ID: (Human) Q8IYS2
Entrez Gene ID: (Human) 90231
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support