Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a transmembrane protein that is preferentially expressed in the nervous system where it controls neuronal cell survival, differentiation into exons and dendrites, and synaptic plasticity. The encoded protein interacts with membrane receptors, cytosolic signaling components, and cytoskeletal proteins, serving as a scaffold that mediates crosstalk between the neurotrophin pathway and several other intracellular signaling pathways. Aberrant expression of this gene is associated with the onset of various neuropsychiatric disorders and neurodegenerative diseases, including Alzheimer's disease. Naturally occurring mutations in this gene are associated with a syndrome characterized by spastic paraplegia, intellectual disability, nystagmus and obesity. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat-rich membrane spanning; Ankyrin repeat-rich membrane-spanning protein; Kinase D-interacting substrate of 220 kDa; kinase D-interacting substrate, 220kDa; MGC163482; truncated KIDINS220
Gene Aliases: 3110039L19Rik; AI194387; AI316525; ARMS; C330002I19Rik; KIAA1250; KIDINS220; mKIAA1250
UniProt ID: (Human) Q9ULH0
Entrez Gene ID: (Human) 57498, (Mouse) 77480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support