Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Immunogen sequence: ESLLATEKVI SSLEKSRDSD KKVVADLMNQ IQELRTSVCE KTETIDTLKQ ELKDINCKYN SALVDREESR VLIKKQEVDI LD
Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.
Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly. Target protein's mouse homolog associates with microtubulew and appears to play a role in mitosis in fibroblasts and in migration in developing neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hKLP2; kinesin-like 7; Kinesin-like protein 2; Kinesin-like protein 7; Kinesin-like protein KIF15; Serologically defined breast cancer antigen NY-BR-62
Gene Aliases: HKLP2; KIF15; KLP2; KNSL7; NY-BR-62
UniProt ID: (Human) Q9NS87
Entrez Gene ID: (Human) 56992
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support