Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: NMEEDRYRLM LSRSNSENIA SNYFRSKWAS QILSTDARKS LTHHNSPPGL SCPLSNNSAI PPTQAPEMPQ PRPFRLESLD IPFTKAKRKK SKSNFGSEPL
Dendrite-specific motor protein which, in association with the Apba1-containing complex (LIN-10-LIN-2-LIN-7 complex), transports vesicles containing N-methyl-D-aspartate (NMDA) receptor subunit NR2B along microtubules. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF17 variant protein; KIF3-related motor protein; Kinesin-like protein KIF17
Gene Aliases: KIAA1405; KIF17; KIF17B; KIF3X; KLP-2; OSM-3
UniProt ID: (Human) Q9P2E2
Entrez Gene ID: (Human) 57576
Molecular Function:
glycosyltransferase
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support