Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HeLa, A-549, Mouse spleen, Mouse testis, Mouse heart, Mouse lung, Mouse brain, Rat spleen. The target is usually found in the following locations: Cytoplasm, Golgi apparatus, cytoskeleton, spindle.
Immunogen sequence: EICNEMVEQM QQREQWCSEH LDTQKELLEE MYEEKLNILK ESLTSFYQEE IQERDEKIEE LEALLQEARQ QSVAHQQSGS ELALRRSQRL AASASTQQLQ EVKAKLQQCK AELNSTTEEL HKYQKMLEPP PSAKPFTIDV DKKLEEGQKN IRLLRTELQK LGESLQSAER ACCHSTGAGK LRQALTTCDD ILIKQDQTLA ELQNNMVLVK LDLRKKAACI AEQYHTVLKL QGQVSAKKRL GTNQENQQPN QQPPGKKPFL RNLLPRTPTC QSSTDCSPYA RILRSRRSPL LKSGPFGKKY
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GG10_2; Kinesin-like protein 174; Kinesin-like protein KIF20A; Mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); Rab6-interacting kinesin-like protein; Rabkinesin-6
Gene Aliases: AA415432; KIF20A; MKLP2; RAB6KIFL
UniProt ID: (Human) O95235, (Mouse) P97329
Entrez Gene ID: (Human) 10112, (Mouse) 19348, (Rat) 361308
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support