Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
KIF21A gene ontology annotations related to this gene include microtubule-based movement.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20052; Kinesin-like protein KIF2; Kinesin-like protein KIF21A; Renal carcinoma antigen NY-REN-62
Gene Aliases: CFEOM1; FEOM1; FEOM3A; KIAA1708; KIF2; KIF21A
UniProt ID: (Human) Q7Z4S6
Entrez Gene ID: (Human) 55605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support