Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VAVADTVREC PPVAGPDGLS KAWGRGGVCT SALVTPTPGS VGGSTGPSAA ASFFIRAMQK LSLASKRKKP
Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF26A variant protein; Kinesin-like protein KIF26A
Gene Aliases: KIAA1236; KIF26A
UniProt ID: (Human) Q9ULI4
Entrez Gene ID: (Human) 26153
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support