Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSESGAASPG ARTRSLKSPK KRATGLQRRR LIPAPLPDTT ALGRKPSLPG QWVDLPPPLA GSLKEPFEIK VYEIDDVERL QRPRPTPREA PTQGLACVST RLRLAERRQQ RLREVQAKHK HLCEELAETQ GRLMLEPGRW LEQFEVDPEL EPESAEYLAA LERATAALEQ CVNLCKAHVM MVTCFDISVA ASAAIPGPQE VDV
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF26A variant protein; Kinesin-like protein KIF26A
Gene Aliases: KIAA1236; KIF26A
UniProt ID: (Human) Q9ULI4
Entrez Gene ID: (Human) 26153
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support