Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GIGTSSPGSL GGSPGFGTGS PGSGSGGGSS PGSDRGVWCE NCNARLVELK RQALRLLLP
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
KIF26B is a member of kinesin-11 family and plays important role in embryogenesis. It is a downstream target of Sall1 and is essential for kidney development. KIF26B is also involved in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinesin-like protein KIF26B
Gene Aliases: KIF26B
UniProt ID: (Human) Q2KJY2
Entrez Gene ID: (Human) 55083
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support