Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
KIF3C is a microtubule-based anterograde translocator for membranous organelles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF3C variant protein; Kinesin-like protein KIF3C
Gene Aliases: KIF3C
UniProt ID: (Human) O14782
Entrez Gene ID: (Human) 3797
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support