Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF5A variant protein; kif5a {ECO:0000312|RGD:1303035}; Kinesin heavy chain isoform 5A; Kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Neuronal kinesin heavy chain; NKHC
Gene Aliases: D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; KIF5A; Kns; mKIAA4086; MY050; NKHC; NKHC1; SPG10
UniProt ID: (Human) Q12840, (Rat) Q6QLM7, (Mouse) P33175
Entrez Gene ID: (Human) 3798, (Pig) 100519957, (Rat) 314906, (Mouse) 16572
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support