Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: VDQCRHRLLM EFDIWYNESF VIPEDMQMAL KPGGSIRPGM VPVNRIVSLG EDDQDKFSQL QQRVLPEGPD SISFYNAKVK IEQKHNYLKT MMGLQQAHR
KIF9 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinesin protein 9; Kinesin-like protein KIF9
Gene Aliases: KIF9
UniProt ID: (Human) Q9HAQ2
Entrez Gene ID: (Human) 64147
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support