Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KIF9 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinesin 9; kinesin protein 9; Kinesin-like protein KIF9
Gene Aliases: KIF9; RGD1565187
UniProt ID: (Human) Q9HAQ2, (Mouse) Q9WV04
Entrez Gene ID: (Human) 64147, (Rat) 501059, (Mouse) 16578
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support