Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ARSAARPAL AQCRALSVDW AGPGSPHGLY LTLQVEHLKE KLISQAQEVS RLRSELGGTD LEKHRDLLMV ENERLRQEMR RCEAELQELR TKPAGPCPGC EHSQESAQLR DKLSQLQLEM AESKGMLSEL NLEVQQKTDR LAEVELRLKD CLAEKAQEEE RLSRRLRDSH ETIASLRAQS PPVKYVIKTV EVESSKTKQA LSESQARNQH LQEQVAMQRQ VLKEMEQQLQ SSHQLTARLR AQI (73-314 aa encoded by BC001211)
KIFC3 belongs to the large superfamily of kinesins, molecular motors that use the energy of ATP hydrolysis to translocate cargoes along microtubules. Members share extensive homology within a globular domain containing the microtubule- and ATP-binding sites and have a coiled-coil stalk domain that mediates oligomerization. Different kinesin family members participate in specific and diverse motile processes, such as cell division, organelle transport, and nuclear movement (Hoang et al., 1998 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686D23201; Kinesin-like protein KIFC3
Gene Aliases: AI325457; BB123200; KIFC3; KRP4
UniProt ID: (Human) Q9BVG8, (Mouse) Q8R3Y4
Entrez Gene ID: (Human) 3801, (Mouse) 16582, (Rat) 307644
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support