Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSMSPTVIIL ACLGFFLDQS VWAHVGGQDK PFCSAWPSAV VPQGGHVTLR CHYRRGFNIF TLYKKDGVPV PELYNRIFWN SFLISPVTPA HAGTYRCRGF HPHSPTEWSA PSNPLVIMVT GLYEKPSLTA RPGPTVRTGE NVTLSCSSQS SFDIYHLSRE GEAHELRLPA VPSINGTFQA DFPLGPATHG ETYRCFGSFH GSPYEWSDAS DPLPVSVTGN PSSSWPSPTE PSFKTGIARH LHAVIRYSVA IILFTILPFF LLHRWCSKKK NAAVMNQEPA GHRTVNREDS DEQDPQEVTY AQLDHCIFTQ RKITGPSQRS KRPSTDTSVC IELPNAEPRA LSPAHEHHSQ ALMGSSRETT ALSQTQLASS NVPAAGI
CD158d / KIR2DL4 is a KIR family member that shares structural features with both activating and inhibitory receptors and may mediate different functions under different circumstances. It contains cytoplasmic ITIM, suggesting inhibitory function, but also transmembrane domain similar to those of activating KIRs. It has been reported that CD158d serves as an inhibitory receptor for peripheral and uterine NK cells, but its ligation with soluble mAbs (unlike immobilized mAbs) results in activation of IFN-gamma secretion. CD158d also binds both membrane form and soluble form of its ligand HLA-G.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD158 antigen-like family member D; CD158d; G9P; Killer cell immunoglobulin-like receptor 2DL4; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; Killer cell inhibitory receptor 103AS; killer Ig receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; KIR-103AS; MHC class I NK cell receptor KIR103AS; natural killer cell inhibitory receptor; natural killer cell receptor; NK cell receptor
Gene Aliases: CD158D; G9P; KIR; KIR-103AS; KIR-2DL4; KIR103; KIR103AS; KIR2DL4
UniProt ID: (Human) Q99706
Entrez Gene ID: (Human) 3805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support