Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: YSTLSGQGRL YDYGQRFVLG MGSSSIELCE REFQRGSLSD SSSFLDTQCD SSVSSSGKQD GYVQFDKASK ASASSSHHSQ SSSQNSDPSR PLQRRMQTHV
The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kin of IRRE-like protein 3; Kin of irregular chiasm-like protein 3; nephrin-like 2; Nephrin-like protein 2
Gene Aliases: KIAA1867; KIRRE; KIRREL3; MRD4; NEPH2; PRO4502; UNQ5923/PRO4502/PRO19814
UniProt ID: (Human) Q8IZU9
Entrez Gene ID: (Human) 84623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support