Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNSLVSWQL LLFLCATHFG EPLEKVASVG NSRPTGQQLE SLGLLAPGEQ SLPCTERKPA ATARLSRRGT SLSPPPESSG SPQQPGLSAP HSRQIPAPQG AVLVQREKDL PNYNWNSFGL RFGKREAAPG NHGRSAGRG (1-138 aa encoded by BC022819 )
This gene is a metastasis suppressor gene that suppresses metastases of melanomas and breast carcinomas without affecting tumorigenicity. The encoded protein may inhibit chemotaxis and invasion and thereby attenuate metastasis in malignant melanomas. Studies suggest a putative role in the regulation of events downstream of cell-matrix adhesion, perhaps involving cytoskeletal reorganization. A protein product of this gene, kisspeptin, stimulates gonadotropin-releasing hormone (GnRH)-induced gonadotropin secretion and regulates the pubertal activation of GnRH neurons. A polymorphism in the terminal exon of this mRNA results in two protein isoforms. An adenosine present at the polymorphic site represents the third position in a stop codon. When the adenosine is absent, a downstream stop codon is utilized and the encoded protein extends for an additional seven amino acid residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 0610012C01Rik; Kiss1 metastasis-suppressor; Kisspeptin-1; malignant melanoma metastasis-suppressor; Metastasis-suppressor KiSS-1; metastin; prepro-kisspeptin
Gene Aliases: HH13; KiSS-1; KISS1; kisspeptin; metastatin; PP5098
UniProt ID: (Human) Q15726, (Mouse) Q6Y4S4
Entrez Gene ID: (Human) 3814, (Mouse) 280287
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support