Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PRTGLSVPAA SGAPYGLLSG YPAMYPAPQY QGHFQLFRGL QGPAPGPATS PSFLS
KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EKLF; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid-specific transcription factor EKLF; Krueppel-like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; truncated EKLF; truncated erythroid Kruppel-like factor 1
Gene Aliases: CDAN4; EKLF; HBFQTL6; INLU; KLF1
UniProt ID: (Human) Q13351
Entrez Gene ID: (Human) 10661
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support