Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcriptional repressor in MAPK/JNK signaling pathway to regulate cellular functions. Overexpression inhibits the transcriptional activities of both the TPA-response element (TRE) and serum response element (SRE). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB and kelch domain-containing protein 6; kelch repeat and BTB (POZ) domain containing 1; Kelch repeat and BTB domain-containing protein 1; kelch-like 31; kelch-like family member 31; Kelch-like protein 31; Kelch-like protein KLHL
Gene Aliases: 9830147P19Rik; bA345L23.2; BKLHD6; D930047P17Rik; KBTBD1; KLHL; KLHL31; RGD1560540
UniProt ID: (Human) Q9H511, (Mouse) Q8BWA5
Entrez Gene ID: (Human) 401265, (Mouse) 244923, (Rat) 315833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support