Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RQGGVNTDK VQCFDPKEDR WSLRSPAPFS QRCLEAVSLE DTIYVMGGLM SKIFTYDPGT DVWGEAAVLP SPVESCGVTV CDGKVHILGG RDDRGESTDK VFTFDPSSGQ VEVQPSLQRC TSSHGCVTII QSLGR (230-363 aa encoded by BC042952)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like 35; kelch-like family member 35; Kelch-like protein 35
Gene Aliases: 2810406K13Rik; KLHL35; RGD1565895
UniProt ID: (Human) Q6PF15, (Mouse) Q9CZ49
Entrez Gene ID: (Human) 283212, (Mouse) 72184, (Rat) 308850
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support