Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLFVIGGARQ GGVNTDKVQC FDPKEDRWSL RSPAPFSQRC LEAVSLEDTI YVMGGLMSKI FT
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like 35; kelch-like family member 35; Kelch-like protein 35
Gene Aliases: KLHL35
UniProt ID: (Human) Q6PF15
Entrez Gene ID: (Human) 283212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support