Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hippostasin; hK11; kallikrein 11; Kallikrein-11; kallikrein-related peptidase 11; Kallikrein11; Serine protease 20; Trypsin-like protease
Gene Aliases: KLK11; PRSS20; TLSP; UNQ649/PRO1279
UniProt ID: (Human) Q9UBX7
Entrez Gene ID: (Human) 11012, (Pig) 110261030, (Rat) 292849
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support