Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ANIQLRSDEE CRQVYPGKIT DNMLCAGTKE GGKDSCEGDS GGPLVCNRTL YGIVSWGDFP CGQPDRPGVY TRVSRYVLWI RETIRKYETQ QQKWLKGPQ
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L4, is expressed in a variety of tissues including prostate, salivary gland, breast, and testis. Its expression is regulated by steroid hormones in breast cancer cell lines may be involved in the pathogenesis and/or progression of breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586J1923; kallikrein 13; Kallikrein-13; kallikrein-like gene 4; Kallikrein-like protein 4; kallikrein-related peptidase 13; KLK-13; KLK-L4
Gene Aliases: KLK-L4; KLK13; KLKL4
UniProt ID: (Human) Q9UKR3
Entrez Gene ID: (Human) 26085
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support