Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFLLLTALQV LAIAMTQSQE DENKIIGGHT CTRSSQPWQA ALLAGPRRRF LCGGALLSGQ WVITAAHCGR PILQVALGKH NLRRWEATQQ VLRVVRQVTH PNYNSRTHDN DLMLLQLQQP ARIGRAVRPI EVTQACASPG TSCRVSGWGT ISSPIARYPA SLQCVNINIS PDEVCQKAYP RTITPGMVCA GVPQGGKDSC QGDSGGPLVC RGQLQGLVSW GMERCALPGY PGVYTNLCKY RSWIEETMRD K
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hK14; Kallikrein L6; Kallikrein-14; Kallikrein-like protein 6; kallikrein-related peptidase 14; KLK-14; KLK-L6
Gene Aliases: KLK-L6; KLK14; KLKL6
UniProt ID: (Human) Q9P0G3
Entrez Gene ID: (Human) 43847
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support