Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACO protease; Kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK-15; prostinogen
Gene Aliases: ACO; HSRNASPH; KLK15
UniProt ID: (Human) Q9H2R5
Entrez Gene ID: (Human) 55554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support