Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MAPSHLSVRE MREDEKPLVL EMLKAGVKDT ENRVALHALT RPPALLLLAA ASSGLR
Probable acetyltransferase that binds the 5'-GGACTACAG-3' sequence of coproporphyrinogen oxidase promoter. Able to activate transcription of a reporter construct in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; N-acetyltransferase 14 (GCN5-related, putative); Probable N-acetyltransferase 14
Gene Aliases: KLP1; NAT14
UniProt ID: (Human) Q8WUY8
Entrez Gene ID: (Human) 57106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support