Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: VTNIFQCIQE KHQRQEILRN CSEKYIMQND NYLKEQILTN KTLKYDVLKN SFQQKKELDS RLIQKNRCHR ENEIVFKVLQ NTGKFSEDHG SCCGVNCYYF TMQKKD
Ly49A is a gene that encodes a protein expressed on the surface of natural killer cells, which is involved in recognizing and eliminating virally infected or cancerous cells. It is located on chromosome 6 and is part of a larger family of genes that encode for similar receptors. The Ly49A protein interacts with a variety of ligands, including certain viruses and tumor cells, and is involved in regulating the activation and function of natural killer cells. Mutations or variations in the Ly49A gene have been associated with various diseases and conditions, including autoimmune disorders and cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KLRA#
Gene Aliases: KLRA1; KLRAP1; Ly-49L; Ly49; LY49L
Entrez Gene ID: (Human) 10748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support