Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQDEERYMT LNVQSKKRSS AQTSQLTFKD YSVTLHWYKI LLGISGTVNG ILTLTLISLI LLVSQGVLLK CQKGSCSNAT QYEDTGDLKV NNGTRRNISN KDLCASRSAD QT (1-111 aa encoded by BC098257)
KLRF1, an activating homodimeric C-type lectin-like receptor (CTLR), is expressed on nearly all natural killer (NK) cells and stimulates their cytoxicity and cytokine release (Kuttruff et al., 2009 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating coreceptor NKp80; C-type lectin domain family 5 member C; CLEC5CMGC119908; killer cell lectin-like receptor F1; Killer cell lectin-like receptor subfamily F member 1; killer cell lectin-like receptor subfamily F, member 1; Lectin-like receptor F1; MGC119907; MGC119909; natural killer cell receptor
Gene Aliases: CLEC5C; KLRF1; ML; NKp80
UniProt ID: (Human) Q9NZS2
Entrez Gene ID: (Human) 51348
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support