Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TIQRVRAEER EQIKTLNNKF ASFIDKVRFL EQQNKVLETK WALLQEQGSR TVRQNLEPLF DSYTSELRRQ LESITTERGR LEAELRNMQD VVEDFKVRYE
This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-75; cytokeratin type II; Cytokeratin-75; hK6hf; K75; keratin 75, type II; Keratin, type II cytoskeletal 75; Keratin-6 hair follicle; Keratin-75; type II keratin-18; Type II keratin-K6hf; Type-II keratin Kb18
Gene Aliases: K6HF; KB18; KRT75; PFB
UniProt ID: (Human) O95678
Entrez Gene ID: (Human) 9119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support