Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: SH-SY5Y
Immunogen sequence: DADINGLRQV LDNLTMEKSD LEMQYETLQE ELMALKKNHK EEMSQLTGQN SGDVNVEINV APGKDLTKTL NDMRQEYEQL IAKNRKDIEN QYETQITQIE HEVSSSGQEV QSSAKEVTQL RHGVQELEIE LQSQLSKKAA LEKSLEDTKN RYCGQLQMI
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-9; cytokeratin 9; Cytokeratin-9; K9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; Keratin, type I cytoskeletal 9; Keratin-9; krt9; Spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9
Gene Aliases: CK-9; EPPK; K9; Krt1-9; KRT9
UniProt ID: (Human) P35527, (Rat) Q8CIS9, (Mouse) Q6RHW0
Entrez Gene ID: (Human) 3857, (Rat) 266717, (Mouse) 107656
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support