Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments, and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif. The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain potassium channel protein-1; IA; MBK1; megencephaly; MGC126782; MGC138385; MKI; potassium (K+) channel protein voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 1; potassium voltage gated channel shaker related subfamily member 1; potassium voltage gated channel, shaker related subfamily, member 1; Potassium voltage-gated channel subfamily A member 1; potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia); RBKI; RCK1; Voltage-gated K(+) channel HuKI; Voltage-gated potassium channel HBK1; Voltage-gated potassium channel subunit Kv1.1
Gene Aliases: AEMK; AI840627; EA1; HBK1; HUK1; Kca1-1; Kcna; KCNA1; Kcpvd; KV1.1; MBK1; mceph; Mk-1; MK1; RBK1; Shak
UniProt ID: (Human) Q09470, (Mouse) P16388, (Rat) P10499
Entrez Gene ID: (Human) 3736, (Mouse) 16485, (Rat) 24520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support