Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total KLK1.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7S nerve growth factor gamma chain; 7S nerve growth factor gamma subunit; Esterase B; Gamma-NGF; glandular kallikrein 1; Glandular kallikrein K1; Glandular kallikrein K3; Glandular kallikrein-7, submandibular/renal; KAL-B; Kallikrein 1 renal/pancreas/salivary; Kallikrein 1, renal/pancreas/salivary; kallikrein 1-like peptidase; Kallikrein 1-related peptidase b3; kallikrein 5; kallikrein 6; kallikrein 7; kallikrein renal/pancreas/salivary; kallikrein serine protease 1; Kallikrein-1; Kallikrein-related protein K1; Kidney/pancreas/salivary gland kallikrein; mGK-3; mGK-6; Nerve growth factor gamma chain; nerve growth factor gamma polypeptide; nerve growth factor, gamma; Pancreatic kallikrein; Proteinase A; PS kallikrein; Renal kallikrein; RGK-1; rGK-7; rK-1; RSKG-7; Tissue kallikrein; Tissue kallikrein-3; Tissue kallikrein-6; True kallikrein; True tissue kallikrein
Gene Aliases: 0610007D04Rik; hK1; Kal; KALA; Klk-1; Klk-3; Klk-6; Klk-7; KLK1; Klk1b3; Klk1b6; Klk1c1; Klk1l; Klk3; Klk5; Klk6; Klk7; KLKR; mGk-3; mGk-6; Ngfg; RATKALA; rGK-1; RGK-7; rK-1; RSKG-7
UniProt ID: (Human) P06870, (Rat) P00758, (Mouse) P15947, (Mouse) A2RTW1, (Rat) P36373
Entrez Gene ID: (Human) 3816, (Rat) 24594, (Mouse) 16612, (Mouse) 18050, (Rat) 24523
Molecular Function: protease serine protease protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support