Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Esterase B; Gamma-NGF; Glandular kallikrein-7, submandibular/renal; Kallikrein 1 renal/pancreas/salivary; Kallikrein 1, renal/pancreas/salivary; kallikrein 1-like peptidase; kallikrein 1-related peptidase b3; kallikrein 5; kallikrein 7; Kallikrein-1; Kallikrein-related protein K1; Nerve growth factor gamma chain; nerve growth factor gamma polypeptide; Pancreatic kallikrein; Proteinase A; PS kallikrein; renal kallikrein; RGK-1; rGK-7; rK-1; RSKG-7; Tissue kallikrein; True kallikrein; True tissue kallikrein
Gene Aliases: Kal; KALA; Klk-1; Klk-7; Klk1; Klk1b3; Klk1c1; Klk1l; Klk5; Klk7; Ngfg; RATKALA; rGK-1; RGK-7; rK-1; RSKG-7
UniProt ID: (Rat) P00758, (Rat) P36373
Entrez Gene ID: (Rat) 24594, (Rat) 24523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support