Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-4240 detects Kallikrein-14 from human samples.
PA1-4240 has been successfully used in Western blot procedures.
PA1-4240 immunizing peptide corresponds to amino acid residues 239-251from human Kallikrein-14.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hK14; Kallikrein L6; Kallikrein-14; Kallikrein-like protein 6; kallikrein-related peptidase 14; KLK-14; KLK-L6
Gene Aliases: KLK-L6; KLK14; KLKL6
UniProt ID: (Human) Q9P0G3
Entrez Gene ID: (Human) 43847
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support