Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glandular kallikrein 2; Glandular kallikrein-1; hGK-1; kallikrein 2, prostatic; Kallikrein-2; kallikrein-related peptidase 2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2
Gene Aliases: hGK-1; hK2; KLK2; KLK2A2
UniProt ID: (Human) P20151
Entrez Gene ID: (Human) 3817
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support