Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TIQLINDHVK KETRGKIVDL VSELKKDVLM VLVNYIYFKA LWEKPFISSR TTPKDFYVDE NTTVRVPMML QDQEHHWYLH DRYLPCSVLR MDYKGDATVF FILPNQGKMR EIEEVLTPEM LMRWNNLLRK RNFYKKLELH LPKFSISGSY VLDQILPRLG FTDLFSKWAD LSGITKQQKL EASKSFHKAT LDVDEAGTEA AAATSFAIKF FSAQTNRHIL RFNRPFLVVI FSTSTQSVLF LGKVVDPTKP; Positive Samples: Mouse pancreas, Rat pancreas
SERPINA4 inhibits human amidolytic and kininogenase activities of tissue kallikrein. The inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kallikrein inhibitor; Kallistatin; Peptidase inhibitor 4; PI-4; protease inhibitor 4 (kallistatin); serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; Serpin A4; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4; sserpin family A member 4; testicular secretory protein Li 22
Gene Aliases: KAL; kallistatin; KLST; KST; PI-4; PI4; SERPINA4
UniProt ID: (Human) P29622
Entrez Gene ID: (Human) 5267, (Rat) 246328
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support