Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; Katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255|HAMAP-Rule:MF_03023}; katanin p60 subunit A 1; Katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255|HAMAP-Rule:MF_03023}; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255|HAMAP-Rule:MF_03023}
Gene Aliases: KATNA1
UniProt ID: (Human) O75449, (Rat) Q6E0V2
Entrez Gene ID: (Human) 11104, (Rat) 292464, (Mouse) 23924
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support