Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
PA1-643 detects kinesin 5B from mouse and human samples. This antibody is specific for kinesin 5B and does not detect other kinesin isotypes.
PA1-643 has been successfully used in Western blot and immunofluorescent procedures. By Western blot, this antibody detects an ~110 kDa protein representing kinesin 5B protein.
The PA1-643 immunizing peptide corresponds to amino acid residues 376-396 from human kinesin 5B. This sequence is 95% and 90% conserved for mouse and rat, respectively. This peptide (Cat. # PEP-198) is available for use in neutralization and control experiments.
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Conventional kinesin heavy chain; epididymis secretory protein Li 61; kif5b {ECO:0000312|EMBL:ABC25059.1}; kinesin 1 (110-120kD); kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; Kinesin-1 heavy chain; U-KHC; Ubiquitous kinesin heavy chain; UKHC
Gene Aliases: AL022807; HEL-S-61; Khc; Khcs; KIF5B; KINH; KNS; KNS1; UKHC
UniProt ID: (Human) P33176, (Mouse) Q61768, (Rat) Q2PQA9
Entrez Gene ID: (Human) 3799, (Mouse) 16573, (Rat) 117550
Molecular Function:
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support