Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
11926-1-AP was generated against N-terminal 300 aa of HMW kininogen. It can bind both HMW and LMW kininogen.
Immunogen sequence: ESQSEEIDC NDKDLFKAVD AALKKYNSQN QSNNQFVLYR ITEATKTVGS DTFYSFKYEI KEGDCPVQSG KTWQDCEYKD AAKAATGECT ATVGKRSSTK FSVATQTCQI TPAEGPVVTA QYDCLGCVHP ISTQSPDLEP ILRHGIQYFN NNTQHSSLFM LNEVKRAQRQ VVAGLNFRMT YSIVQTNCSK ENFLFLTPDC KSLWNGDTGE CTDNAYIDIQ LRIASFSQNC DIYPGKDFVQ PPTKICVGCP RDIPTNSPEL EETLTHTITK LNAENNATFY FKIDNVKKAR VQVVAGKKYF IDFVARETTC SKESNEELTE SCETKKLGQS LDCNAEVYVV PWEKKIYPTV NCQPLG (20-374 aa encoded by BC060039)
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2-thiol proteinase inhibitor; bradykinin; Fitzgerald factor; High molecular weight kininogen; HMWK; Kininogen-1; Williams-Fitzgerald-Flaujeac factor
Gene Aliases: BDK; BK; KNG; KNG1
UniProt ID: (Human) P01042
Entrez Gene ID: (Human) 3827
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support