Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
In Western blots, reacts with full length Kininogen (aa 21-661) and several forms of the light chain starting at residue 389, and does not react with the heavy chain. In Western blots, no cross-reactivity with recombinant mouse (rm) Cystatin A, B, C, E/M, recombinant human (rh) Cystatin D, F, S, SA, SN, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-kininigen; Kininogen-1; L-kininogen
Gene Aliases: Kng; Kng1
UniProt ID: (Mouse) O08677
Entrez Gene ID: (Mouse) 16644
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support